Transfection delivery system, dynamic delivery system, TransIT-X2™
Lieferant: MIRUS
TransIT-X2™ Dynamic Delivery System is a polymeric system that efficiently delivers both DNA and RNA out of the endosome and into the cytoplasm overcoming a critical barrier to nucleic acid delivery.
- Efficiency – exceptional broad spectrum transfection
- Versatility – cutting edge delivery of plasmid DNA and siRNA
- Technology – non liposomal, polymeric delivery
TransIT-X2® Dynamic Delivery System is in 80% ethanol and should be stored tightly capped at −20 °C.
TransIT-X2 can be used for DNA delivery, siRNA or miRNA delivery or simultaneous co-transfection of DNA and siRNA or miRNA. Transfections with TransIT-X2 do not require medium changes and can be carried out in serum-containing medium.
Bestellinformation: TransIT-X2® dynamic delivery system is available in 0,3, 0,75, 1,5 ml, 5×1,5 ml (5 pack) and 10×1,5 ml, (10 pack) configurations.