Expression system, CHOgro®
Lieferant: MIRUS
MIR-6260
MIRUMIR-6260EA 981 EUR
MIRUMIR-6260
Expression system, CHOgro®
Transfektionsreagenzien
CHOgro® offers a high titre transient transfection system for suspension CHO cells.
  • Enables high protein titres with simple workflow
  • Quick adaptation to CHO cell lineages
  • High density growth with minimal cell clumping post transfection
  • No commercial license required; animal-origin-free
Lieferumfang: Kit contains CHOgro® expression medium, 2 L; TransIT-PRO® transfection reagent, 1 ml; CHOgro® complex formation solution, 100 ml; Poloxamer 188 solution,100 ml and L-glutamine, 100 ml. All components can also be purchased separately.