Transfection reagent, TransIT®-Keratinocyte
Lieferant: MIRUS
MIR2800 MIR2804 MIR2805 MIR2806
MIRUMIR2800EA 666 EUR
MIRUMIR2800 MIRUMIR2804 MIRUMIR2805 MIRUMIR2806
Transfection reagent, TransIT®-Keratinocyte
Transfektionsreagenzien
TransIT®-Keratinocyte transfection reagent is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in primary and immortalised keratinocytes.
  • High transfection efficiency (20 to 30%)
  • Suitable for both transient and stable transfection
  • Low cellular toxicity: Maintain cell density and reduce experimental bias
Transfection with TransIT®-Keratinocyte reagent does not require medium changes and can be carried out in serum-containing medium.