Transfection reagent, TransIT®-Keratinocyte
Lieferant: MIRUS
MIR2800
MIR2804
MIR2805
MIR2806
MIRUMIR2800EA
666
EUR
MIRUMIR2800
MIRUMIR2804
MIRUMIR2805
MIRUMIR2806
Transfection reagent, TransIT®-Keratinocyte
Transfektionsreagenzien
TransIT®-Keratinocyte transfection reagent is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in primary and immortalised keratinocytes.
- High transfection efficiency (20 to 30%)
- Suitable for both transient and stable transfection
- Low cellular toxicity: Maintain cell density and reduce experimental bias
Transfection with TransIT®-Keratinocyte reagent does not require medium changes and can be carried out in serum-containing medium.