Nucleic acid labeling kits, Label IT®
Lieferant: MIRUS
Label IT® nucleic acid labeling kits are suitable for direct non-enzymatic labeling of DNA and RNA. Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications.
- Easily and consistently control the labeling reactions
- Achieve high sensitivity with optimally labeled DNA and RNA
- Do not impact hybridisation performance
All Label IT® reagent kits use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Lieferumfang: Each kit contains a Label IT® reagent, Label IT® reconstitution solution, 10X labeling buffer and solutions.