G418 Disulfat 50 mg/ml in Wasser

Lieferant: MIRUS

Synonyme: G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat

MIR5920
MIRUMIR5920EA 136 EUR
MIRUMIR5920
G418 Disulfat 50 mg/ml in Wasser
G418 disulphate
Formel: C₂₀H₄₄N₄O₁₈S₂
MDL: MFCD00058314
CAS-Nummer: 108321-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR