Hygromycin B 50 mg/ml in PBS

Lieferant: MIRUS

MIR5930
MIRUMIR5930EA 292 EUR
MIRUMIR5930
Hygromycin B 50 mg/ml in PBS
Hygromycin B
Formel: C₂₀H₃₇N₃O₁₃
MDL: MFCD06795479
CAS-Nummer: 31282-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR