Puromycin Dihydrochlorid 10 mg/ml water

Lieferant: MIRUS

Synonyme: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

MIR5940
MIRUMIR5940EA 430 EUR
MIRUMIR5940
Puromycin Dihydrochlorid 10 mg/ml water
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Molecular Weight: 544.44 g/mol
MDL: MFCD00012691
CAS-Nummer: 58-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR