Human IgG1 expression control
Lieferant: MIRUS
MIR6250
MIRUMIR6250EA 558 EUR
MIRUMIR6250
Human IgG1 expression control
Transfektionsreagenzien
Human IgG1 Expression Control is a heavy and light chain mixture ideal for verifying antibody expression in mammalian cells.
  • Equimolar ratios of human IgG1 heavy and light chain constructs designed using DNA2.0 IP-Free vector technology
  • Verify human IgG1 expression as a control using ELISA or other methods
  • Suitable for use with any large scale antibody expression system
Lieferumfang: Supplied ready-to-use at 1,0 mg/ml concentration.