TransduceIT Reagent is an aqueous solution of hexadimethrine bromide, a cationic polymer, that is shown to greatly enhance retroviral transduction and transgene expression in both dividing and quiescent mammalian cells. Provided as 1mL vial. 1 * 1 mL
Lieferant: MIRUS
TransduceIT Reagent is an aqueous solution of hexadimethrine bromide, a cationic polymer, that is shown to greatly enhance retroviral transduction and transgene expression in both dividing and quiescent mammalian cells. Provided as 1mL vial. 1 * 1 mL