Intracellular nucleic acid localisation kits, Label IT® Tracker™
Lieferant: MIRUS
Label IT® Tracker™ is designed for efficient, direct labeling of plasmid DNA for in vitro and in vivo tracking experiments.
- Efficiently label and visualise the DNA of choice
- Easily and consistently control the labeling reactions
- Optimal visualisation of DNA in cells
- Monitor both subcellular localization and reporter transgene expression following transfection of labeled (plasmid) DNA
Lieferumfang: Each kit contains Label IT® Tracker™ reagent, Label IT® reconstitution solution and 10X labeling buffer, sufficient to label 50 to 200 µg of plasmid DNA.