Plasmid delivery controls, Label IT®
Lieferant: MIRUS
MIR7907
MIR7906
MIR7905
MIR7904
MIRUMIR7907EA
967
EUR
MIRUMIR7907
MIRUMIR7906
MIRUMIR7905
MIRUMIR7904
Plasmid delivery controls, Label IT®
Vektoren, Plasmide und Bibliotheken
Fluorescently labelled plasmids ideal for in vitro and in vivo tracking.
- For in vivo delivery
- Easily visualise transfected cells and assess delivery efficiency using fluorescent microscopy
- Suitable for co-delivery experiments with functional plasmids
Lieferumfang: Supplied as a ready to use 0,5 mg/ml concentrate.