Conductive tips, MBP
Lieferant: Thermo Fisher Scientific
904-013
MLBP904-013EA 1380 EUR
MLBP904-013
Conductive tips, MBP
Pipettenspitzen
Filtered, conductive tip style, liquid sensing, Econopacks.
  • For use with: Tecan™ Freedom EVO™ LiHa Head, Tecan™ Genesis™
  • Ideal for automated liquid handling systems
  • Non sterile, polypropylene
Perkin-Elmer is a registered trademark of Perkin-Elmer Corporation. ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. EconoPak is a trademark of Molecular BioProducts. Tecan is a registered trademark of Tecan AG. Cavro is a registered trademark of Scientific Instruments, Inc. MultiPROBE and Packard are registered trademarks of Packard Instrument Company, Inc. Thermo Orion is a registered trademark of Thermo Orion Inc.