CellXpress.ai Automated Cell Culture System
Lieferant: Molecular Devices
GU04162050 GU04162040 GU04162030 GU04162020 GU04161020 GU04161010 GU04130005 GU04130004 GU04106008 GU04106007 GU04106006 GU04106005 GU04106004 GU04106003 GF4110002 GF4110001 CELLXPRESS_AI
MLDVGU04162050EA 0 EUR
MLDVGU04162050 MLDVGU04162040 MLDVGU04162030 MLDVGU04162020 MLDVGU04161020 MLDVGU04161010 MLDVGU04130005 MLDVGU04130004 MLDVGU04106008 MLDVGU04106007 MLDVGU04106006 MLDVGU04106005 MLDVGU04106004 MLDVGU04106003 MLDVGF4110002 MLDVGF4110001 MLDVCELLXPRESS_AI
CellXpress.ai Automated Cell Culture System
Automated Cell Culture Systems
AI-driven cell culture innovation hub that gives your team total control over demanding cell culture feeding and passaging schedules—eliminating time in the lab while maintaining a 24/7 schedule for growing and scaling multiple stem cell lines, spheroids, or organoids.
  • automate your cell culture process
  • improve your screening workflows
  • develop reliable, reproducible assays
This revolutionary solution can give your lab the confidence in experimental outcomes to make key decisions sooner, achieve milestones faster, and get to clinic earlier—with lowerMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable