CloneMatrix
Lieferant: Molecular Devices
K8510
K8530
MLDVK8510EA
249
EUR
MLDVK8510
MLDVK8530
CloneMatrix
Zellkulturmedien
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.