Industrielle Abdeckbänder, 301E
Lieferant: 3M
301E-18MM 301E-24MM 301E-36MM 301E-48MM
MMMA301E-18MMEA 258 EUR
MMMA301E-18MM MMMA301E-24MM MMMA301E-36MM MMMA301E-48MM
Industrielle Abdeckbänder, 301E
Klebebänder
301E masking tape is a thick crepe paper tape for industrial applications that offers good water resistance.
  • Withstands surface temperatures up to 225 °F/107 °C for 30 minutes
  • Resistant to solvents and moisture
  • Easy and neat detachment of many heating systems after curing
  • Designed for indoor use
  • Tensile strength: 95 [Newton/25 mm]
  • Crepe paper back material
It provides good holding power that resists lifting or curling, making it suitable for air drying and oven curing of paint and lacquer.
Using this proprietary solvent-free adhesive, the tape sticks instantly to most surfaces, including metal, rubber, glass and plastic, as well as EPDM-type rubber moldings while removing cleanly in one piece. Our high-strength crepe-paper backing conforms to irregular surfaces, resists bleed through from most industrial paint systems, improves visibility during the paint masking process, and helps you achieve good paint lines.