Fit Test Adapters
Lieferant: 3M
601
MMMA601EA 223 EUR
MMMA601
Fit Test Adapters
Atemschutzmasken
For an alternative method to sharing a probed fit testing respirator, use the 3M™ quantitative fit test adapter. The adaptor kits allow users to fit test with a personal 3M™ Respirator, rather than using a shared probed fit test respirator.
  • Adapter allows fit testing with an individual's personal 3M™ respirator
  • Alternative to sharing a probed fit testing respirator
  • Quick and easy to fit
The 3M™ Quantitative fit test adapters are designed for use with 3M™ half and full-face respirators with bayonet-style filter attachments.