Filters, DT
Lieferant: 3M
7100260622
MMMA7100260622EA 13.5 EUR
MMMA7100260622
Filters, DT
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
For effective protection against a range of particulates, gases, and vapours, or combination hazards, use 3M™ filters DT series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ powered air respirator system PF-600E+ series.
  • Includes specifications to provide protection against particulate hazards, gas, and vapour hazards, or a combination of both
  • Filters feature a DIN 40mm threaded connection
  • Please note: DT-Series filters were previously known as Pro2000 filters
The DT series filters are available in a range of specifications, and are designed for use with 3M reusable and powered air respirators with DIN 40 filter ports. This filter series is compatible with 3M™ full facepiece Reusable Respirator FF-600 Series, 3M™ Powered Air Respirator System PF-600-ASB Series, 3M™ Half facepiece reusable respirator HF-300 series, 3M™ full facepiece reusable respirator FF-300 series, and 3M™ powered air respirator PF-600E.
Recommended applications include use in demolition, grinding, and metalworking.