Doppelseitiges Schaumklebeband 9515W
Lieferant: 3M
9515W12
MMMA9515W12EA 9880 EUR
MMMA9515W12
Doppelseitiges Schaumklebeband 9515W
Klebebänder
3M™ Double coated foam tape 9515W is a white, 1,5 mm thick, double coated polyethylene foam tape with acrylic adhesive.
  • Closed-cell double coated polyethylene foam with acrylic adhesive
  • Mount and join a wide range of materials
  • Suitable for different surfaces
The acrylic adhesive provides good initial tack and adhesion to a wide variety of surfaces. Intended applications include assembly of displays (POS), lightweight emblems, name plates, etc.