Partikelfilter der Serie D3000, Secure Click™
Lieferant: 3M
D3125 D3135 D3138
111-2606EA 356 EUR
111-2606 111-2607 MMMAD3138
Partikelfilter der Serie D3000, Secure Click™
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
3M™ Secure Click™ D3000 Series Particulate Filters feature a new dual flow design for easier breathing.
  • New dual flow filters allow for easier breathing
  • Protects against particulates with nuisance level organic vapour/acid gas relief
  • Protection against ozone up to 10× occupational exposure limit
  • Quick, simple connection; simply align connections and push until an audible click is heard
  • Use with 3M™ Secure Click™ Reusable Respirators 800 Series
  • Swept-back design allows an enhanced field of view and comfort
  • Use for protection in hazardous environments