Scotch-Brite™ 770 Cellulosic Sponge Laminate
Lieferant: 3M
SCH770
MMMASCH770EA 28.2 EUR
MMMASCH770
Scotch-Brite™ 770 Cellulosic Sponge Laminate
Wischtücher Schwämme
Scotch-Brite™ Cellulosic sponge laminate 770 is a dual-function cleaning tool made from a combination of a non-woven tablecloth containing very light abrasive particles to a pure cellulose sponge.
  • Great cleaning performance
  • High absorption power
  • Does not scratch even the most sensitive surfaces
  • Works with hot water or diluted bleach
  • Excellent hydrophilic properties
Remove grease and food without leaving scratches with Scotch-Brite™ Cellulosic Sponge Laminate 770. Made from a combination of a non-woven tablecloth containing very light abrasiveMore Product Information