SecureFit™ 200 Safety Glasses
Lieferant: 3M
SF201AFP SF202AFP SF203AFP
MMMASF201AFPEA 9.7 EUR
MMMASF201AFP MMMASF202AFP MMMASF203AFP
SecureFit™ 200 Safety Glasses
Schutzbrillen
3M™ SecureFit™ 200 series are lightweight safety glasses with 3M™ Pressure Diffusion Temple Technology. The glasses feature a wraparound design that’s secure and comfortable. They are available in various lens tints.
  • Economical, lightweight (18 g) safety glasses range
  • Innovative SecureFit™ Pressure Diffusion Temple technology features slotted ribs that provide comfortable pressure equalisation
  • Temples flex and self-adjust to the size of wearer's head
  • Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Wraparound design improves the security of fit and prevents slipping
  • Flat and flexible temples combine easily with earmuffs
  • 99,9% UV protection
  • Easy to clean
For lightweight comfort at an affordable price, use 3M™ SecureFit™ Safety Glasses 200 series. These safety glasses weigh just 18 g and offer a comfortable fit. The 3M™ Pressure Diffusion Temple Technology allows the temple touchpoints to naturally adjust to an individual’s head size. Wide, flexible temple arms over the ears provide solid side protection and work well with earmuffs. The wraparound design gives the user a wide field of vision and helps to reduce slippage from head movements.
Foam gasket available to help limit eye exposure to debris. Prescription insert available.
Standards: EN 166:2001.