3M™ SecureFit™ Safety glasses 600 series are semi-rimless glasses with 3M™ pressure diffusion temple technology and fixed-length temple arms over the ears.
- Robust semi-rimless safety glasses with dual-lenses provide optimal optical performance
- The Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalisation
- Temples flex and self-adjust to the size of wearer's head
- Scotchgard™ Anti-Fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
- Polarised and green IR filters available
- Wraparound design helps a secure fit and prevents slipping
- Foam gasket available to help limit eye exposure to debris
For optimal optical performance and good field of vision, use the SecureFit™ Safety Glasses 600 Series. These innovative and stylish semi-rimless, grey framed safety glasses have two separate lenses for a good field of vision and wide, fixed-length temple arms over the ears for a close, comfortable fit and solid side protection. They feature Pressure Diffusion Temple Technology to help diffuse pressure over the ear and soft nose pads to enhance frame comfort across a diverse workforce. The wraparound design helps provide a secure fit and prevents slipping. These glasses combine well with ear muffs.