Oil Sorbent Pillows
Lieferant: 3M
T3016
MMMAT3016EA 195 EUR
MMMAT3016
Oil Sorbent Pillows
Absorptionsmaterial
3M™ Oil sorbent pillows are open mesh bags designed with a scrim that encases particulate sorbent. Excellent for handling large quantities of light oil in confined spaces, they are UV-resistant and will not deteriorate or sink.
  • Ideal for absorbing oil from areas where water may also be present
  • Open mesh bags are tear-resistant and feature a high capacity for holding liquid
  • Ideal for placing at the source of a leak
Being hydrophobic repelling water, they are ideal for absorbing oil from areas where water may also be present.