Rapid readout biological indicator 1295 for vaporised hydrogen peroxide sterilisation, Attest™
Lieferant: 3M MEDICAL
1295
MMME1295EA 1560 EUR
MMME1295
Rapid readout biological indicator 1295 for vaporised hydrogen peroxide sterilisation, Attest™
Sterilisationsindikatoren
3M™ Attest™ Rapid Readout Biological Indicators 1295 for vaporised hydrogen peroxide (VH2O2) sterilisation provide faster results.
  • Biological indicator provides results in just 4 hours (or less)
  • Positive results can be obtain in 15 minutes allowing rapid corrective action, before surgical sets are sent to the operated room
  • Ideal for load monitoring
The 3M™ Attest 490H Auto reader can be remotely accessed using a web browser, allowing digital record keeping. The 490H Auto readers can also be linked to Track&trace systems.
When incubated and read automatically in the user-friendly 3M™ Attest™ Auto-reader 490H, Rapid read-out technology provides biological indicator results in just 4 hours - allowing you to have the information you need while there's still time to act. Simplify your workflow and provide safety for your staff and your patients.
For use with STERRAD® 100S, STERRAD® NX® (Standard and Advanced cycles), STERRAD® 100 NX® (Standard, FLEX, Express and Duo cycles), and STERIS® AMSCO® VPRO® maX (Lumen, Non Lumen, and Flexible cycles) system vaporised hydrogen peroxide sterilisers.
Cap colour: Pink