Attest™ 390 Auto-Reader for Steam
Lieferant: 3M MEDICAL
M390
MMMEM390EA 4550 EUR
MMMEM390
Attest™ 390 Auto-Reader for Steam
Sterilisationsindikatoren
This auto-reader for steam, 390, is designed to incubate and automatically read the biological indicators for steam at 60 °C for a final fluorescent result.
  • Reliable and accurate
  • Features advanced electronics and a backlit LCD panel
  • Intelligent display with time-to-result countdown feature
  • Audible alarm sounds for improved monitoring
  • Display can be viewed remotely using your smartphone or PC
Colour: Light grey/charcoal