Spreaders, Petrifilm™
Lieferant: 3M Food Safety
6481 6425 6498 6496 6497
710-0776EA 40.7 EUR
710-0776 MMMP6425 MMMP6498 MMMP6496 MMMP6497
Spreaders, Petrifilm™
Schaber Impfösen und Impfnadeln
This 3M™ Petrifilm™ Spreader is an easy-to-use tool designed to quickly and evenly distribute inoculum within the 3M™ Petrifilm™ Plate.
  • Design easily spreads inoculum over uniform area
  • The spreader is reusable and does not contact plating surface or sample
This spreader is compatible with the following 3M™ Petrifilm™ Plates: Aerobic count plates, Coliform count plates, rapid Coliform count plates, Enterobacteriaceae count plates, E. coli/Coliform count plates, aqua Coliform count plates and aqua heterotrophic count plates. Once the sample has beenMore Product Information
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Environmentally Preferable