Allergen protein ELISA kits

Lieferant: 3M Food Safety

E96BZL E96CAS E96CHW E96CRU E96EGG E96FSH E96HZL E96MAC E96MLK E96MOL E96MUS E96PEC E96PNT E96PST E96SES E96SOY E96ALM
MMMPE96BZLEA 1030 EUR
MMMPE96BZL MMMPE96CAS MMMPE96CHW MMMPE96CRU MMMPE96EGG MMMPE96FSH MMMPE96HZL MMMPE96MAC MMMPE96MLK MMMPE96MOL MMMPE96MUS MMMPE96PEC MMMPE96PNT MMMPE96PST MMMPE96SES MMMPE96SOY MMMPE96ALM
Allergen protein ELISA kits
Tests ELISAs

The allergen protein ELISA Kit is intended for the detection of mollusk proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.


  • Designed to detect both processed and unprocessed target allergen proteins
  • Test can accommodate environmental swab, CIP rinse water and food samples
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time
  • 50 minutes test time
  • Storage: 2 to 8 °C


These kits utilise a sandwich ELISA. The proteins present in the sample react with the antibody, which have been adsorbed to the surface of polystyrene microtiter wells. After the removal of unbound proteins by washing, antibodies conjugated with horseradish peroxidase (HRP) are added. These enzyme-labelled antibodies form complexes with the previously bound protein. Following a second washing step, the enzyme bound to the immunosorbent is detected by the addition of a chromogenic substrate, 3,3’,5,5’-tetramethylbenzidine (TMB). The colour development from this enzymatic reaction varies directly with the concentration of protein in the sample tested; thus, the absorbance, at 450 nm, is a measure of the concentration of protein in the test sample. The quantity of protein in the test sample can be extrapolated from the standard curve, constructed from standards of known concentration, and adjusted to consider the sample dilution.

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
Target protein Allergen: Brazil nut Allergen: Bovine casein Allergen: Cashew Allergen: Crustacean Allergen: Egg white Allergen: Fish Allergen: Hazelnut Allergen: Macadamia Allergen: Bovine total milk Allergen: Mollusk Allergen: Mustard Allergen: Pecan Allergen: Peanut Allergen: Pistachio Allergen: Sesame Allergen: Soy Allergen: Almond
Reaktivität der primären Antikörper
Testtyp Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich Sandwich
Wirt
Konjugat-ELISA HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP HRP
Format Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated Pre-coated
Erkennungsmethode Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm) Colorimetric (450 nm)
Nachweisgrenze Protein thresholdd: 1 ppm Protein threshold: 0,5 ppm Protein threshold: 0,9 ppm Protein thresholdd: 2 ppm Protein threshold: 0,5 ppm Protein thresholdd: 1 ppm Protein thresholdd: 1 ppm Protein threshold: 0,3 ppm Protein thresholdd: 1 ppm Protein thresholdd: 1 ppm Protein threshold: 1,5 ppm Protein threshold: 0,7 ppm Protein thresholdd: 2 ppm Protein thresholdd: 1 ppm Protein thresholdd: 2 ppm Protein thresholdd: 2 ppm Protein thresholdd: 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR