Molecular detection matrix control
Lieferant: 3M Food Safety
MDMC96NA
MMMPMDMC96NAEA 808 EUR
MMMPMDMC96NA
Molecular detection matrix control
Tests Immunologische Tests
Kit contains 96 tests and is used with the 3M™ Molecular Detection System to check for inhibition of the matrix sample during validation periods, when adopting the 3M™ method or testing new or unknown matrices.
  • Verify that new or unknown matrices are not inhibitory to amplification
  • Ready-to-use and pre-dispensed reagents
  • Only two transfer steps after enrichment
  • Closed-tube system reduces risk of amplicon contamination
  • Designed to work with 3M™ proprietary enrichment broths to promote growth