Molecular detection instrument and accessory kit
Lieferant: 3M Food Safety
MDS100EU
MMMPMDS100EUEA
30190
EUR
MMMPMDS100EU
Molecular detection instrument and accessory kit
Tests
Immunologische Tests
The 3M™ Molecular detection instrument comes with a pluggable heater, software, power cord, USB cable and a basic accessory kit.
- Smaller footprint than a standard notebook computer that requires significantly less space than the market leading rapid methods
- Robust instrument that is solid state, portable with no need for recalibration, requires minimal maintenance, and provides automatic diagnostics at start up
- Powerful software that allows operation of up to four instruments simultaneously from one computer
The basic accessory kit includes a chill block insert, chill block tray, speed loader tray, cap/decap tool lysis, cap/decap tool reagent, and empty lysis and reagent trays.