Magnetic stirrer for heating mantles
Lieferant: MOHR
KM-MR3000
MOHRKM-MR3000EA 619 EUR
MOHRKM-MR3000
Magnetic stirrer for heating mantles
Rührer Magnetrührer
This magnetic stirrer with controller for standard heating mantles from 500 to 4000 ml, for stirring the medium in a round glass flask.
  • Safety class I
  • Having rotational frequency upto 1600 U/min
The heating mantle must be positioned on the top of the stirrer and the controllable heat together with the stir feature ensures equal heat transfer up to 450 °C.
Standards: IP 33 protection class.