Digital timer, 579CE
Lieferant: MOINEAU
0579CE
MOIN0579CEEA 68.5 EUR
MOIN0579CE
Digital timer, 579CE
Timer Timer, digital
This digital timer features an illuminated key, a memory and an alarm volume adjustment.
  • Timing capacity: 99 min 59 seconds
  • Counts up and down
  • Alarm: 1 minute, three different volumes (silent, normal, loud)
  • Illuminated key: green (counting) / red (alarm)
  • Large display is easy to read
  • Can be attached to a metal surface thanks to its magentic back
Lieferumfang: Supplied with 2 AAA batteries.