Digital timer, two channels, 580CE
Lieferant: MOINEAU
0580CE
MOIN0580CEEA 95.2 EUR
MOIN0580CE
Digital timer, two channels, 580CE
Timer Timer, digital
Digital timer with two channels and two display lines.
  • Timing capacity: 100 hours
  • Counts down and up
  • Memory for both channels
  • Adjustable alarm volume
  • Acoustic and visual alarm
Weight: 85 g
Lieferumfang: Supplied with stand, clamp and magnet. Batteries (2× AAA) need to be ordered separately.