Digital thermo-hygrometer
Lieferant: MOINEAU
9222AT
MOIN9222ATEA 122 EUR
MOIN9222AT
Digital thermo-hygrometer
Hygrometer Thermohygrometer, digital
This small thermo-hygrometer can be used for outdoor and indoor measurements.
  • Large and easy-to-read display
  • Storage of Min/Max values
  • Steel probe with 3 m cable
Lieferumfang: Supplied with wall-mount and batteries (2× AAA).