2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3)
Lieferant: Molekula
|
Achtung
|
Synonyme:
dTTP-Na3
10019137-100MG
MOLE10019137-100MGEA
394
EUR
MOLE10019137-100MG
2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3)
2'-Deoxythymidine-5'-triphosphate trisodium salt (dTTP-Na3)
Formel:
C₁₀H₁₄N₂O₁₄P₃·3Na Molecular Weight: 548,114 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00080338 CAS-Nummer: 3624-46-2 EINECS: 222-826-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...