3-Methoxyphenylboronsäure
Lieferant: Molekula
|
Achtung
|
Synonyme:
(3-Methoxyphenyl)metaborsäure, Dihydroxy(3-methoxyphenyl)boran, 3-Methoxybenzolboronsäure
10126175-1G
MOLE10126175-1GEA
0
EUR
MOLE10126175-1G
3-Methoxyphenylboronsäure
3-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ Molecular Weight: 151,96 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161359 CAS-Nummer: 10365-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...