5-Fluor-5'-deoxyuridin (Doxifluridin)

Lieferant: Molekula

10130089-25MG
MOLE10130089-25MGEA 0 EUR
MOLE10130089-25MG
5-Fluor-5'-deoxyuridin (Doxifluridin)
5-Fluor-5'-deoxyuridin (Doxifluridin)
Formel: C₉H₁₁FN₂O₅
MDL: MFCD00866530
CAS-Nummer: 3094-09-5
EINECS: 221-440-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR