(4-Nitrophenyl)-β-D-glucopyranosid
Lieferant: Molekula
Synonyme:
(4-Nitrophenyl)-β-D-glucosid
10159337-5G
MOLE10159337-5GEA
291
EUR
MOLE10159337-5G
(4-Nitrophenyl)-β-D-glucopyranosid
(4-Nitrophenyl)-β-D-glucopyranosid
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301.25 g/mol |
MDL:
MFCD00006593 CAS-Nummer: 2492-87-7 EINECS: 219-661-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...