3-(3-Indolyl)propionsäure
Lieferant: Molekula
Synonyme:
3-(Indol-3-yl)propionsäure, 3-Indolpropionsäure, 3-Indol-3-propionsäure
10186726-100G
MOLE10186726-100GEA
218
EUR
MOLE10186726-100G
3-(3-Indolyl)propionsäure
3-(3-Indolyl)propionsäure
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,21 g/mol Schmelzpunkt: 129…132 °C Dichte: 1,356 g/cm³ (20 °C) |
MDL:
MFCD00005660 CAS-Nummer: 830-96-6 EINECS: 212-600-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...