2,5-Dichlor-4-nitroanilin

Lieferant: Molekula
Gefahr

10250092-5G
MOLE10250092-5GEA 0 EUR
MOLE10250092-5G
2,5-Dichlor-4-nitroanilin
2,5-Dichlor-4-nitroanilin
Formel: C₆H₄Cl₂N₂O₂
Molecular Weight: 207,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025157
CAS-Nummer: 6627-34-5
EINECS: 229-591-5
UN: 2237
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR