Methyl-3,3-dimethoxypropionat
Lieferant: Molekula
Synonyme:
Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate
10262793-25G
MOLE10262793-25GEA
0
EUR
MOLE10262793-25G
Methyl-3,3-dimethoxypropionat
Methyl-3,3-dimethoxypropionat
Formel:
C₆H₁₂O₄ Molecular Weight: 148,16 g/mol |
MDL:
MFCD00010650 CAS-Nummer: 7424-91-1 EINECS: 231-055-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...