1,1'-Bis(diphenylphosphino)ferrocen
Lieferant: Molekula
Synonyme:
DPPF, 1,1-Bis(diphenylphosphino)ferrocene
10332118-1G
MOLE10332118-1GEA
51.8
EUR
MOLE10332118-1G
1,1'-Bis(diphenylphosphino)ferrocen
1,1'-Bis(diphenylphosphino)ferrocen
Formel:
C₃₄H₂₈FeP₂ Molecular Weight: 554,39 g/mol |
MDL:
MFCD00001422 CAS-Nummer: 12150-46-8 EINECS: 430-420-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...