3-Amino-5-methylisoxazol

Lieferant: Molekula

Synonyme: 5-Methylisoxazol-3-ylamin, 5-Methylisoxazol-3-amin

10347617-100G
MOLE10347617-100GEA 0 EUR
MOLE10347617-100G
3-Amino-5-methylisoxazol
3-Amino-5-methylisoxazole
Formel: C₄H₆N₂O
Molecular Weight: 98.1 g/mol
Siedepunkt: 194
Schmelzpunkt: 56…60 °C
MDL: MFCD00003155
CAS-Nummer: 1072-67-9
EINECS: 214-013-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR