(2-Bromphenyl)essigsäure
Lieferant: Molekula
Synonyme:
2-(2-Bromophenyl)acetic acid
10435778-25G
MOLE10435778-25GEA
0
EUR
MOLE10435778-25G
(2-Bromphenyl)essigsäure
(2-Bromphenyl)essigsäure
Formel:
(C₆H₄Br)CH₂COOH Molecular Weight: 215,05 g/mol Siedepunkt: 308 °C (1013 hPa) Schmelzpunkt: 107,5 °C |
MDL:
MFCD00004314 CAS-Nummer: 18698-97-0 EINECS: 242-509-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...