L(-)-Altrose

Lieferant: Molekula

Synonyme: L-Altrose, L-Altropyranose

10692892-25MG
MOLE10692892-25MGEA 0 EUR
MOLE10692892-25MG
L(-)-Altrose
L(-)-Altrose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
MDL: MFCD09880209
CAS-Nummer: 1949-88-8
EINECS: 217-764-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR