N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)

Lieferant: Molekula

Synonyme: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA

10783994-5G
MOLE10783994-5GEA 0 EUR
MOLE10783994-5G
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Formel: C₉H₁₈F₃NOSi
Molecular Weight: 241,33 g/mol
MDL: MFCD00009671
CAS-Nummer: 77377-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR