2-Chlor-4,6-dimethoxy-1,3,5-triazin

Lieferant: Molekula

Synonyme: 2-Chlor-4,6-dimethoxy-s-triazin

10809468-25G
MOLE10809468-25GEA 100 EUR
MOLE10809468-25G
2-Chlor-4,6-dimethoxy-1,3,5-triazin
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Formel: C₅H₆ClN₃O₂
Molecular Weight: 175,57 g/mol
MDL: MFCD00075607
CAS-Nummer: 3140-73-6
EINECS: 221-541-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR