1H-1,2,4-Triazol-3-thiol

Lieferant: Molekula

Synonyme: 3-Mercapto-1H-1,2,4-triazol, 3-Mercapto-1,2,4-triazol

10821503-25G
MOLE10821503-25GEA 44.9 EUR
MOLE10821503-25G
1H-1,2,4-Triazol-3-thiol
1H-1,2,4-Triazol-3-thiol
Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
Schmelzpunkt: 217…220 °C
MDL: MFCD00005229
CAS-Nummer: 3179-31-5
EINECS: 221-656-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR