(4-Nitrophenyl)acetonitril
Lieferant: Molekula
Synonyme:
4-Nitrobenzylcyanid
10922744-25G
MOLE10922744-25GEA
61
EUR
MOLE10922744-25G
(4-Nitrophenyl)acetonitril
(4-Nitrophenyl)acetonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Siedepunkt: 336 °C (1013 hPa) Schmelzpunkt: 108…113 °C |
MDL:
MFCD00007372 CAS-Nummer: 555-21-5 EINECS: 209-085-0 UN: 3439 ADR: 6.1,III Merck Index: 12,06688 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...