5-Methylchinoxalin

Lieferant: Molekula
Achtung

10923708-5G
MOLE10923708-5GEA 285 EUR
MOLE10923708-5G
5-Methylchinoxalin
5-Methylquinoxaline
Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012335
CAS-Nummer: 13708-12-8
EINECS: 237-246-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR